Learn More
Meso Scale Discovery U-PLEX Human Eotaxin-3 Assay, K251UEK-4, 384-well format, single-analyte, LLOD 7.3 pg/mL, dynamic range 7.3-21,400 pg/mL, 25 plates
SDP
Catalog No. NC2895632
Encompass
The U-PLEX Human Eotaxin-3 Assay is a highly sensitive immunoassay designed for precise quantitative detection of human Eotaxin-3 in various biological samples including plasma, serum, and cell culture supernatants. It leverages the flexibility of the U-PLEX platform, facilitating single-analyte assays on 384-well plates with opportunities for multiplex integration.
Catalog No. NC2895632 Supplier Meso Scale Discovery Supplier No. K251UEK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human Eotaxin-3 Assay is a highly sensitive immunoassay designed for precise quantitative detection of human Eotaxin-3 in various biological samples including plasma, serum, and cell culture supernatants. It leverages the flexibility of the U-PLEX platform, facilitating single-analyte assays on 384-well plates with opportunities for multiplex integration.