Learn More
Meso Scale Discovery U-PLEX Human Eotaxin ASY
SDP
Catalog No. 5000082019
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin Assay Kit is designed for the quantitative measurement of human Eotaxin in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for detection of human Eotaxin with no cross-reactivity to other proteins It includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent The kit can perform up to 80 tests across two plates
Catalog No. 50-000-82019 Supplier Meso Scale Discovery Supplier No. K151UDK1
The Meso Scale Discovery U-PLEX Human Eotaxin Assay Kit is designed for the quantitative measurement of human Eotaxin in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for detection of human Eotaxin with no cross-reactivity to other proteins It includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent The kit can perform up to 80 tests across two plates