Learn More
Meso Scale Discovery U-PLEX Human Eotaxin ASYQP
SDP
Catalog No. 5000082020
Encompass
The MSD U-PLEX Human Eotaxin Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of eotaxin (CCL11) in human biological samples This singleplex format allows focused analysis of this key chemokine involved in eosinophil recruitment and allergic responses The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods
Catalog No. 50-000-82020 Supplier Meso Scale Discovery Supplier No. K151UDK22
The MSD U-PLEX Human Eotaxin Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of eotaxin (CCL11) in human biological samples This singleplex format allows focused analysis of this key chemokine involved in eosinophil recruitment and allergic responses The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods