Learn More
Meso Scale Discovery U-PLEX Human EPO ASY
SDP
Catalog No. 5000079540
Encompass
The Meso Scale Discovery U-PLEX Human EPO Assay Kit is an electrochemiluminescent immunoassay for measuring erythropoietin (EPO) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation of EPO with detection limits from 0 14 to 400 mIU/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range allowing high-throughput EPO quantitation from small samples
Catalog No. 50-000-79540 Supplier Meso Scale Discovery Supplier No. K151VXK1
The Meso Scale Discovery U-PLEX Human EPO Assay Kit is an electrochemiluminescent immunoassay for measuring erythropoietin (EPO) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation of EPO with detection limits from 0 14 to 400 mIU/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range allowing high-throughput EPO quantitation from small samples