Learn More
Meso Scale Discovery U-PLEX Human Factor VII ASY
SDP
Catalog No. 5000079922
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is designed to quantitatively measure Factor VII levels in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 1500 ng/mL This product enables high-throughput quantification of Factor VII with a short 3 5 hour protocol
Catalog No. 50-000-79922 Supplier Meso Scale Discovery Supplier No. K151N9K1
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is designed to quantitatively measure Factor VII levels in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 1500 ng/mL This product enables high-throughput quantification of Factor VII with a short 3 5 hour protocol