Learn More
Meso Scale Discovery U-PLEX Human Factor VII ASY
SDP
Catalog No. 5000082327
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is a multiplex immunoassay that measures Factor VII levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay can be used for biomarker research related to coagulation disorders
Catalog No. 50-000-82327 Supplier Meso Scale Discovery Supplier No. K151N9K2
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is a multiplex immunoassay that measures Factor VII levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay can be used for biomarker research related to coagulation disorders