Learn More
Meso Scale Discovery U-PLEX Human Factor VII ASYQP
SDP
Catalog No. 5000079601
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit measures Factor VII in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and buffers Results are available in 3 5 hours using an MSD instrument This assay offers a rapid sensitive method for measuring Factor VII with a wide dynamic range
Catalog No. 50-000-79601 Supplier Meso Scale Discovery Supplier No. K151N9K21
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit measures Factor VII in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and buffers Results are available in 3 5 hours using an MSD instrument This assay offers a rapid sensitive method for measuring Factor VII with a wide dynamic range