Learn More
Meso Scale Discovery U-PLEX Human Fas (soluble) Assay, 96-well plate, 0.22 pg/mL LLOD, Dynamic Range: 0.22 - 1600 pg/mL, for serum and plasma, Mouse Monoclonal antibodies, Research use only
SDP
Catalog No. NC3239539
Encompass
The U-PLEX Human Fas (soluble) Assay by MESO SCALE DISCOVERY is a sophisticated immunoassay designed for the quantitative detection of soluble Fas in human serum and plasma. This assay utilizes validated components of the U-PLEX platform, simplifying the development of sensitive assays suitable for future multiplexing within Immuno-Oncology analytes. Intended solely for research purposes, this product ensures reliable results in oncological and immunological applications.
  • Designed for quantitative detection of soluble Fas.
  • Suitable for serum and plasma samples.
  • Dynamic range of 0.22 - 1,600 pg/mL.
  • Sensitivity level of 0.22 pg/mL.
  • Compatible with multiple instruments.
  • Incorporates both capture and detection antibodies from mouse monoclonal sources.
  • Part of Immuno-Oncology Group 1 multiplex compatibility.
  • Multi-component storage conditions may apply.
Catalog No. NC3239539 Supplier Meso Scale Discovery Supplier No. K151AQCK2
The U-PLEX Human Fas (soluble) Assay by MESO SCALE DISCOVERY is a sophisticated immunoassay designed for the quantitative detection of soluble Fas in human serum and plasma. This assay utilizes validated components of the U-PLEX platform, simplifying the development of sensitive assays suitable for future multiplexing within Immuno-Oncology analytes. Intended solely for research purposes, this product ensures reliable results in oncological and immunological applications.
  • Designed for quantitative detection of soluble Fas.
  • Suitable for serum and plasma samples.
  • Dynamic range of 0.22 - 1,600 pg/mL.
  • Sensitivity level of 0.22 pg/mL.
  • Compatible with multiple instruments.
  • Incorporates both capture and detection antibodies from mouse monoclonal sources.
  • Part of Immuno-Oncology Group 1 multiplex compatibility.
  • Multi-component storage conditions may apply.