Learn More
Meso Scale Discovery U-PLEX Human FGF-23 ASYQP
SDP
Catalog No. 5000082421
Encompass
The U-PLEX Human FGF-23 Assay (K1516EK) is a multiplex electrochemiluminescence immunoassay designed to quantitatively measure fibroblast growth factor 23 (FGF-23) levels in human serum plasma and cell culture samples This research-grade assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements FGF-23 is a key biomarker for phosphate homeostasis and bone metabolism research making this assay valuable for studies in chronic kidney disease bone disorders and metabolic research applications
Catalog No. 50-000-82421 Supplier Meso Scale Discovery Supplier No. K1516EK21
The U-PLEX Human FGF-23 Assay (K1516EK) is a multiplex electrochemiluminescence immunoassay designed to quantitatively measure fibroblast growth factor 23 (FGF-23) levels in human serum plasma and cell culture samples This research-grade assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements FGF-23 is a key biomarker for phosphate homeostasis and bone metabolism research making this assay valuable for studies in chronic kidney disease bone disorders and metabolic research applications