Learn More
Meso Scale Discovery U-PLEX Human FGF basic ASY
SDP
Catalog No. 5000081923
Encompass
The Meso Scale Discovery U-PLEX Human FGF Basic Assay is a multiplex immunoassay kit for quantifying human FGF basic in various biological matrices It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 series The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags and a calibrator with recombinant FGF basic
Catalog No. 50-000-81923 Supplier Meso Scale Discovery Supplier No. K151X7K1
The Meso Scale Discovery U-PLEX Human FGF Basic Assay is a multiplex immunoassay kit for quantifying human FGF basic in various biological matrices It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 series The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags and a calibrator with recombinant FGF basic