Learn More
Meso Scale Discovery U-PLEX Human FGF basic ASY
SDP
Catalog No. 5000081924
Encompass
The Meso Scale Discovery U-PLEX Human FGF Basic Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human FGF basic in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use
Catalog No. 50-000-81924 Supplier Meso Scale Discovery Supplier No. K151X7K2
The Meso Scale Discovery U-PLEX Human FGF Basic Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human FGF basic in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use