Learn More
Meso Scale Discovery U-PLEX Human Fractalkine ASY
SDP
Catalog No. 5000081861
Encompass
The Meso Scale Discovery U-PLEX Human Fractalkine Assay is a multiplex immunoassay kit for detecting and quantifying fractalkine in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent technology and includes plates with capture antibodies detection antibodies calibrators controls buffers and reagents for 96 tests The assay detects fractalkine from 19 5 to 10 000 pg/mL takes 3 5 hours and requires an MSD instrument It shows no cross-reactivity with other cytokines and chemokines
Catalog No. 50-000-81861 Supplier Meso Scale Discovery Supplier No. K151VCK2
The Meso Scale Discovery U-PLEX Human Fractalkine Assay is a multiplex immunoassay kit for detecting and quantifying fractalkine in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent technology and includes plates with capture antibodies detection antibodies calibrators controls buffers and reagents for 96 tests The assay detects fractalkine from 19 5 to 10 000 pg/mL takes 3 5 hours and requires an MSD instrument It shows no cross-reactivity with other cytokines and chemokines