Learn More
Meso Scale Discovery U-PLEX Human FSH ASY384-well
SDP
Catalog No. 5000078634
Encompass
The Meso Scale Discovery U-PLEX Human FSH 384-Well Assay Kit is a multiplex immunoassay that measures Follicle Stimulating Hormone (FSH) levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides a sensitive accurate and reproducible way to quantify FSH levels across a broad dynamic range It is available in a 384-well plate format includes all necessary reagents and can process hundreds of samples in just a few hours The assay has high specificity no cross-reactivity and requires only a small sample volume
Catalog No. 50-000-78634 Supplier Meso Scale Discovery Supplier No. K2516GK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FSH 384-Well Assay Kit is a multiplex immunoassay that measures Follicle Stimulating Hormone (FSH) levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides a sensitive accurate and reproducible way to quantify FSH levels across a broad dynamic range It is available in a 384-well plate format includes all necessary reagents and can process hundreds of samples in just a few hours The assay has high specificity no cross-reactivity and requires only a small sample volume