Learn More
Meso Scale Discovery U-PLEX Human G-CSF ASY 25 PL
SDP
Catalog No. 5000079544
Encompass
The Meso Scale Discovery U-PLEX Human G-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference This assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure G-CSF concentrations precisely
Catalog No. 50-000-79544 Supplier Meso Scale Discovery Supplier No. K151VGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human G-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference This assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure G-CSF concentrations precisely