Learn More
Meso Scale Discovery U-PLEX Human G-CSF ASY
SDP
Catalog No. 5000081395
Encompass
The Meso Scale Discovery U-PLEX Human G-CSF Assay is a multiplex immunoassay kit that allows simultaneous measurement of Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure G-CSF concentrations
Catalog No. 50-000-81395 Supplier Meso Scale Discovery Supplier No. K151VGK1
The Meso Scale Discovery U-PLEX Human G-CSF Assay is a multiplex immunoassay kit that allows simultaneous measurement of Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure G-CSF concentrations