Learn More
Meso Scale Discovery U-PLEX Human Galectin-9 ASY
SDP
Catalog No. 5000078801
Encompass
The Meso Scale Discovery Galectin-9 Kit is an electrochemiluminescent immunoassay for measuring galectin-9 in various biological samples It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 1 3 pg/mL with a range of 1 5-1000 pg/mL The assay time is 3 5 hours for 96 samples enabling accurate galectin-9 quantification for research
Catalog No. 50-000-78801 Supplier Meso Scale Discovery Supplier No. K151AGWK1
The Meso Scale Discovery Galectin-9 Kit is an electrochemiluminescent immunoassay for measuring galectin-9 in various biological samples It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 1 3 pg/mL with a range of 1 5-1000 pg/mL The assay time is 3 5 hours for 96 samples enabling accurate galectin-9 quantification for research