Learn More
Meso Scale Discovery U-PLEX Human Galectin-9 ASY
SDP
Catalog No. 5000079521
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a multiplex immunoassay kit for quantifying galectin-9 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and minimal matrix interference The assay requires 50 L or less sample volume and runs in under 2 5 hours It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid reproducible measurement of galectin-9 crucial in immune regulation and cancer progression
Catalog No. 50-000-79521 Supplier Meso Scale Discovery Supplier No. K151AGWK2
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a multiplex immunoassay kit for quantifying galectin-9 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and minimal matrix interference The assay requires 50 L or less sample volume and runs in under 2 5 hours It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid reproducible measurement of galectin-9 crucial in immune regulation and cancer progression