Learn More
Meso Scale Discovery U-PLEX Human Galectin-9 ASYQP
SDP
Catalog No. 5000078802
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is an electrochemiluminescent immunoassay for quantifying Galectin-9 in human serum plasma and cell culture supernatants It uses MSD s SECTOR technology is sensitive reproducible and works with common sample matrices The assay measures Galectin-9 from 0 05 to 1000 ng/mL with a 50 L sample The U-PLEX format allows customization and multiplexing with other assays for simultaneous biomarker analysis providing high-performance sensitive and reproducible quantification of Galectin-9
Catalog No. 50-000-78802 Supplier Meso Scale Discovery Supplier No. K151AGWK21
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is an electrochemiluminescent immunoassay for quantifying Galectin-9 in human serum plasma and cell culture supernatants It uses MSD s SECTOR technology is sensitive reproducible and works with common sample matrices The assay measures Galectin-9 from 0 05 to 1000 ng/mL with a 50 L sample The U-PLEX format allows customization and multiplexing with other assays for simultaneous biomarker analysis providing high-performance sensitive and reproducible quantification of Galectin-9