Learn More
Meso Scale Discovery U-PLEX Human Galectin-9 ASYQP
SDP
Catalog No. 5000080471
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a sensitive immunoassay for measuring human Galectin-9 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology delivering accurate results in 3 5 hours The U-PLEX platform allows for measuring multiple biomarkers in a 25-50 L sample It has a sensitivity with an LLOQ of 19 5 pg/mL and a dynamic range up to 10 000 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for Galectin-9 analysis
Catalog No. 50-000-80471 Supplier Meso Scale Discovery Supplier No. K151AGWK22
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is a sensitive immunoassay for measuring human Galectin-9 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology delivering accurate results in 3 5 hours The U-PLEX platform allows for measuring multiple biomarkers in a 25-50 L sample It has a sensitivity with an LLOQ of 19 5 pg/mL and a dynamic range up to 10 000 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for Galectin-9 analysis