Learn More
Meso Scale Discovery U-PLEX Human GITR/TNFRSF18 ASY
SDP
Catalog No. 5000081810
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology offering sensitive and reproducible results with a detection range of 19 to 100 000 pg/mL The assay requires 50 L of sample per well and is compatible with MSD s SECTOR Imager platforms enabling high-throughput multiplexed quantification of GITR levels for research applications
Catalog No. 50-000-81810 Supplier Meso Scale Discovery Supplier No. K151Y7K1
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology offering sensitive and reproducible results with a detection range of 19 to 100 000 pg/mL The assay requires 50 L of sample per well and is compatible with MSD s SECTOR Imager platforms enabling high-throughput multiplexed quantification of GITR levels for research applications