Learn More
Meso Scale Discovery U-PLEX Human GITR/TNFRSF18 ASY
SDP
Catalog No. 5000081841
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology offering sensitive results with a detection range of 19 to 100 000 pg/mL The assay requires 50 L of sample per well and is compatible with MSD s SECTOR Imager platforms The kit includes a plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals
Catalog No. 50-000-81841 Supplier Meso Scale Discovery Supplier No. K151Y7K2
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITR in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology offering sensitive results with a detection range of 19 to 100 000 pg/mL The assay requires 50 L of sample per well and is compatible with MSD s SECTOR Imager platforms The kit includes a plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals