Learn More
Meso Scale Discovery U-PLEX Human GITRL/TNFSF18 ASY
SDP
Catalog No. 5000078514
Encompass
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITRL in various biological matrices It uses MSD s electrochemiluminescent technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay is automatable with a 3 5-hour turnaround and includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and measuring sample concentrations
Catalog No. 50-000-78514 Supplier Meso Scale Discovery Supplier No. K151S7K1
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay kit for detecting and quantifying human GITRL in various biological matrices It uses MSD s electrochemiluminescent technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay is automatable with a 3 5-hour turnaround and includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and measuring sample concentrations