Learn More
Meso Scale Discovery U-PLEX Human GM-CSF 384-Well Assay, 4-pack
SDP
Catalog No. NC3359511
Encompass
The U-PLEX Human GM-CSF 384-Well Assay is designed for precise measurement of human Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) in a single-analyte format. This assay utilizes the U-PLEX platform, providing validated components for the rapid and straightforward development of high-quality immunoassays. It is suitable for use on 384-well plates and offers flexibility for multiplexing GM-CSF with other analytes through U-PLEX Custom Biomarker, Metabolic, or Immuno-Oncology Group 1 (human) Assays.
  • Measures human GM-CSF
  • 384-well assay format
  • Single-analyte assay type
  • Utilizes U-PLEX platform
  • Compatible with multiplexing
  • Includes biotin-conjugated antibodies
  • Components like MSD calibrators, controls, and diluents
  • Research use only
Catalog No. NC3359511 Supplier Meso Scale Discovery Supplier No. K251UMK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human GM-CSF 384-Well Assay is designed for precise measurement of human Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) in a single-analyte format. This assay utilizes the U-PLEX platform, providing validated components for the rapid and straightforward development of high-quality immunoassays. It is suitable for use on 384-well plates and offers flexibility for multiplexing GM-CSF with other analytes through U-PLEX Custom Biomarker, Metabolic, or Immuno-Oncology Group 1 (human) Assays.
  • Measures human GM-CSF
  • 384-well assay format
  • Single-analyte assay type
  • Utilizes U-PLEX platform
  • Compatible with multiplexing
  • Includes biotin-conjugated antibodies
  • Components like MSD calibrators, controls, and diluents
  • Research use only