Learn More
Meso Scale Discovery U-PLEX Human GM-CSF Assay, 96-well SECTOR plates, Electro-chemiluminescence detection, LLOD 0.12 pg/mL, Dynamic range 0.12-9,400 pg/mL, Sample types: Serum, Plasma, Cell Culture Supernatant
SDP
Catalog No. NC3660048
Encompass
U-PLEX Human GM-CSF Assay, 96-well SECTOR plates, Electro-chemiluminescence detection, LLOD 0.12 pg/mL, Dynamic range 0.12-9,400 pg/mL, Sample types: Serum, Plasma, Cell Culture Supernatant
Catalog No. NC3660048 Supplier Meso Scale Discovery Supplier No. K151UMK4
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human GM-CSF Assay, 96-well SECTOR plates, Electro-chemiluminescence detection, LLOD 0.12 pg/mL, Dynamic range 0.12-9,400 pg/mL, Sample types: Serum, Plasma, Cell Culture Supernatant