Learn More
Meso Scale Discovery U-PLEX Human GRO-α Assay, 96-well plate, Human, Serum/Plasma/Cell Culture Supernatant, LLOD 0.25 pg/mL, Dynamic Range 0.25 - 2500 pg/mL
SDP
Catalog No. NC2822010
Encompass
U-PLEX Human GRO-α Assay, 96-well plate, Human, Serum/Plasma/Cell Culture Supernatant, LLOD 0.25 pg/mL, Dynamic Range 0.25 - 2500 pg/mL
Catalog No. NC2822010 Supplier Meso Scale Discovery Supplier No. K151UXK2
U-PLEX Human GRO-α Assay, 96-well plate, Human, Serum/Plasma/Cell Culture Supernatant, LLOD 0.25 pg/mL, Dynamic Range 0.25 - 2500 pg/mL