Learn More
Meso Scale Discovery U-PLEX Human GRO- ASY
SDP
Catalog No. 5000080425
Encompass
The Meso Scale Discovery U-PLEX Human GRO-alpha Assay Kit is a multiplex immunoassay for measuring GRO-alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less sample volume it can be automated on MSD platforms The kit includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curve generation and assay validation It allows simultaneous measurement of GRO-alpha and other biomarkers with high reproducibility
Catalog No. 50-000-80425 Supplier Meso Scale Discovery Supplier No. K151UXK1
The Meso Scale Discovery U-PLEX Human GRO-alpha Assay Kit is a multiplex immunoassay for measuring GRO-alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less sample volume it can be automated on MSD platforms The kit includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curve generation and assay validation It allows simultaneous measurement of GRO-alpha and other biomarkers with high reproducibility