Learn More
Meso Scale Discovery U-PLEX Human GRO-α 384-Well Assay, Immunoassay, Analyte: GRO-α, Plate Format: 384-well, LLOD: 0.25 pg/mL, Dynamic Range: 0.25 - 2500 pg/mL, Sample Types: Plasma, Serum, Cell Culture Supernatant
SDP
Catalog No. NC3433995
Encompass
The U-PLEX Human GRO-α 384-Well Assay is a validated immunoassay kit designed for accurate quantitative measurement of human GRO-α. It leverages the U-PLEX platform for high sensitivity and reproducibility in research applications, making it suitable for various scientific studies. The kit operates in a 384-well plate format, facilitating high-throughput analysis and efficient sample processing.
  • Validated platform for rapid creation of high-quality assays
  • Precisely measures human GRO-α in a single-analyte format
  • Offers a low limit of detection for accurate quantification
  • Validated for use with plasma, serum, and cell culture supernatant
  • Optimized for efficient processing of multiple samples
Catalog No. NC3433995 Supplier Meso Scale Discovery Supplier No. K251UXK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human GRO-α 384-Well Assay is a validated immunoassay kit designed for accurate quantitative measurement of human GRO-α. It leverages the U-PLEX platform for high sensitivity and reproducibility in research applications, making it suitable for various scientific studies. The kit operates in a 384-well plate format, facilitating high-throughput analysis and efficient sample processing.
  • Validated platform for rapid creation of high-quality assays
  • Precisely measures human GRO-α in a single-analyte format
  • Offers a low limit of detection for accurate quantification
  • Validated for use with plasma, serum, and cell culture supernatant
  • Optimized for efficient processing of multiple samples