Learn More
Meso Scale Discovery U-PLEX Human HAVCR2/TIM-3 ASY
SDP
Catalog No. 5000080190
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human TIM-3 and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible analysis of TIM-3 levels alongside other biomarkers
Catalog No. 50-000-80190 Supplier Meso Scale Discovery Supplier No. K151E8K1
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human TIM-3 and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible analysis of TIM-3 levels alongside other biomarkers