Learn More
Meso Scale Discovery U-PLEX Human HAVCR2/TIM-3 ASY
SDP
Catalog No. 5000081642
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit for detecting and quantifying human TIM-3 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators Results are analyzed with MSD s QuickPlex SQ 120 instrument This assay efficiently measures TIM-3 important in immune response regulation
Catalog No. 50-000-81642 Supplier Meso Scale Discovery Supplier No. K151E8K2
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit for detecting and quantifying human TIM-3 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators Results are analyzed with MSD s QuickPlex SQ 120 instrument This assay efficiently measures TIM-3 important in immune response regulation