Learn More
Meso Scale Discovery U-PLEX Human HVEM/TNFRSF14 ASY
SDP
Catalog No. 5000080445
Encompass
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human HVEM in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for human HVEM with a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL The assay requires 50 L of sample per well and can be read on MSD s SECTOR Imager instrument platforms This assay provides a rapid accurate method for measuring levels of the important immune checkpoint protein HVEM
Catalog No. 50-000-80445 Supplier Meso Scale Discovery Supplier No. K151AGXK1
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human HVEM in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for human HVEM with a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL The assay requires 50 L of sample per well and can be read on MSD s SECTOR Imager instrument platforms This assay provides a rapid accurate method for measuring levels of the important immune checkpoint protein HVEM