Learn More
Meso Scale Discovery U-PLEX Human I-309 ASY 25 PL
SDP
Catalog No. 5000079677
Encompass
The Meso Scale Discovery U-PLEX Human I-309 Assay is a multiplex immunoassay kit for measuring human cytokine I-309 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrator solutions and assay diluent The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL enabling high-throughput sensitive and reproducible quantification of human I-309
Catalog No. 50-000-79677 Supplier Meso Scale Discovery Supplier No. K151UYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human I-309 Assay is a multiplex immunoassay kit for measuring human cytokine I-309 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrator solutions and assay diluent The assay sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL enabling high-throughput sensitive and reproducible quantification of human I-309