Learn More
Meso Scale Discovery U-PLEX Human I-309 ASY
SDP
Catalog No. 5000080410
Encompass
The Meso Scale Discovery U-PLEX Human I-309 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human cytokine I-309 in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well or 384-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve This assay allows sensitive accurate and rapid quantification of I-309 levels with as little as 25-50 L of sample
Catalog No. 50-000-80410 Supplier Meso Scale Discovery Supplier No. K151UYK1
The Meso Scale Discovery U-PLEX Human I-309 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human cytokine I-309 in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well or 384-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve This assay allows sensitive accurate and rapid quantification of I-309 levels with as little as 25-50 L of sample