Learn More
Meso Scale Discovery U-PLEX Human I-309 ASY384-well
SDP
Catalog No. 5000079415
Encompass
The Meso Scale Discovery U-PLEX Human I-309/384-Well Assay is a 384-well electrochemiluminescent immunoassay for measuring human I-309 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and controls It has a dynamic range of 0 61-2500 pg/mL and requires 25 L sample per well enabling high-throughput quantification of I-309 a key chemokine in immune regulation
Catalog No. 50-000-79415 Supplier Meso Scale Discovery Supplier No. K251UYK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human I-309/384-Well Assay is a 384-well electrochemiluminescent immunoassay for measuring human I-309 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and controls It has a dynamic range of 0 61-2500 pg/mL and requires 25 L sample per well enabling high-throughput quantification of I-309 a key chemokine in immune regulation