Learn More
Meso Scale Discovery U-PLEX Human I-309 ASYQP
SDP
Catalog No. 5000079005
Encompass
The U-PLEX Human I-309 Assay (K151UYK) is a multiplex immunoassay platform by Meso Scale Discovery for quantitative measurement of I-309 (CCL1) chemokine levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible scalable testing with superior precision and reproducibility compared to traditional ELISA methods making it ideal for research applications in immunology inflammation studies and biomarker discovery
Catalog No. 50-000-79005 Supplier Meso Scale Discovery Supplier No. K151UYK21
The U-PLEX Human I-309 Assay (K151UYK) is a multiplex immunoassay platform by Meso Scale Discovery for quantitative measurement of I-309 (CCL1) chemokine levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible scalable testing with superior precision and reproducibility compared to traditional ELISA methods making it ideal for research applications in immunology inflammation studies and biomarker discovery