Learn More
Meso Scale Discovery U-PLEX Human I-TAC Assay 5 plates
SDP
Catalog No. NC2875041
Encompass
The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human I-TAC Assay has the essential components for an assay that measures I-TAC in a single-analyte format. For information on multiplexing I-TAC with other analytes, please see U-PLEX Custom Biomarker Group 1 (human) Assays or U-PLEX Custom Immuno-Oncology Group 1 (human) Assays.
Catalog No. NC2875041 Supplier Meso Scale Discovery Supplier No. K151UWK2
The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human I-TAC Assay has the essential components for an assay that measures I-TAC in a single-analyte format. For information on multiplexing I-TAC with other analytes, please see U-PLEX Custom Biomarker Group 1 (human) Assays or U-PLEX Custom Immuno-Oncology Group 1 (human) Assays.