Learn More
Meso Scale Discovery U-PLEX Human I-TAC ASY 25 PL
SDP
Catalog No. 5000080375
Encompass
The U-PLEX Human I-TAC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human interferon-inducible T cell alpha chemoattractant (I-TAC) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80375 Supplier Meso Scale Discovery Supplier No. K151UWK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human I-TAC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human interferon-inducible T cell alpha chemoattractant (I-TAC) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL