Learn More
Meso Scale Discovery U-PLEX Human I-TAC ASY
SDP
Catalog No. 5000079120
Encompass
The U-PLEX Human I-TAC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and rapid way to quantify I-TAC levels with high precision and accuracy The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals With a broad dynamic range and low sample volume requirement this assay kit enables efficient analysis of I-TAC levels in research studies
Catalog No. 50-000-79120 Supplier Meso Scale Discovery Supplier No. K151UWK1
The U-PLEX Human I-TAC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and rapid way to quantify I-TAC levels with high precision and accuracy The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals With a broad dynamic range and low sample volume requirement this assay kit enables efficient analysis of I-TAC levels in research studies