Learn More
Meso Scale Discovery U-PLEX Human I-TAC ASY384-well
SDP
Catalog No. 5000080701
Encompass
The U-PLEX Human I-TAC Assay from Meso Scale Discovery is a multiplex immunoassay that measures Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours using an MSD instrument This assay allows for the rapid accurate quantification of I-TAC
Catalog No. 50-000-80701 Supplier Meso Scale Discovery Supplier No. K251UWK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human I-TAC Assay from Meso Scale Discovery is a multiplex immunoassay that measures Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours using an MSD instrument This assay allows for the rapid accurate quantification of I-TAC