Learn More
Meso Scale Discovery U-PLEX Human ICAM-1 ASY
SDP
Catalog No. 5000078746
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed to quantitatively measure Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for sample dilution The assay has a detection range of 78-50 000 pg/mL and exhibits no cross-reactivity with other adhesion molecules This product can aid in understanding ICAM-1 levels related to inflammatory and immune responses
Catalog No. 50-000-78746 Supplier Meso Scale Discovery Supplier No. K151E9K1
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed to quantitatively measure Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for sample dilution The assay has a detection range of 78-50 000 pg/mL and exhibits no cross-reactivity with other adhesion molecules This product can aid in understanding ICAM-1 levels related to inflammatory and immune responses