Learn More
Meso Scale Discovery U-PLEX Human ICAM-1 ASY
SDP
Catalog No. 5000081641
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of ICAM-1 with high sensitivity and a broad dynamic range This product can help researchers study cell adhesion molecules and inflammation biomarkers The kit includes a 96-well plate calibrators diluent detection antibody read buffer wash buffer plate seals and an instruction booklet with assay protocol
Catalog No. 50-000-81641 Supplier Meso Scale Discovery Supplier No. K151E9K2
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of ICAM-1 with high sensitivity and a broad dynamic range This product can help researchers study cell adhesion molecules and inflammation biomarkers The kit includes a 96-well plate calibrators diluent detection antibody read buffer wash buffer plate seals and an instruction booklet with assay protocol