Learn More
Meso Scale Discovery U-PLEX Human ICAM-1 ASYQP
SDP
Catalog No. 5000078532
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed to quantitatively measure Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method This multiplex assay kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG reagents and calibrators and controls to generate a standard curve and validate assay performance The sensitive assay has a lower limit of detection of 0 05 ng/mL and an upper limit of 1000 ng/mL for ICAM-1 The kit includes sufficient reagents for 1 x 96-well plate when run as singleplex
Catalog No. 50-000-78532 Supplier Meso Scale Discovery Supplier No. K151E9K21
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed to quantitatively measure Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method This multiplex assay kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG reagents and calibrators and controls to generate a standard curve and validate assay performance The sensitive assay has a lower limit of detection of 0 05 ng/mL and an upper limit of 1000 ng/mL for ICAM-1 The kit includes sufficient reagents for 1 x 96-well plate when run as singleplex