Learn More
Meso Scale Discovery U-PLEX Human ICAM-1 ASYQP
SDP
Catalog No. 5000080193
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and a U-PLEX linker to capture and detect ICAM-1 in samples The kit includes a 96-well plate calibrator detection antibody read buffer 10X wash buffer sample diluent and plate sealers The assay has a typical sensitivity of 0 23 pg/mL and an upper limit of quantification of 1500 pg/mL for serum/plasma samples
Catalog No. 50-000-80193 Supplier Meso Scale Discovery Supplier No. K151E9K22
The Meso Scale Discovery U-PLEX Human ICAM-1 Assay Kit is designed for the quantitative measurement of human Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and a U-PLEX linker to capture and detect ICAM-1 in samples The kit includes a 96-well plate calibrator detection antibody read buffer 10X wash buffer sample diluent and plate sealers The assay has a typical sensitivity of 0 23 pg/mL and an upper limit of quantification of 1500 pg/mL for serum/plasma samples