Learn More
Meso Scale Discovery U-PLEX Human ICOS ASY 25 PL
SDP
Catalog No. 5000079085
Encompass
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay kit for quantifying human ICOS in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies for human ICOS detection antibodies calibrators controls diluents wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for human ICOS
Catalog No. 50-000-79085 Supplier Meso Scale Discovery Supplier No. K151AHBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay kit for quantifying human ICOS in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies for human ICOS detection antibodies calibrators controls diluents wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for human ICOS