Learn More
Meso Scale Discovery U-PLEX Human ICOS ASY
SDP
Catalog No. 5000079084
Encompass
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human ICOS in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for human ICOS detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of human ICOS levels
Catalog No. 50-000-79084 Supplier Meso Scale Discovery Supplier No. K151AHBK2
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human ICOS in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for human ICOS detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of human ICOS levels