Learn More
Meso Scale Discovery U-PLEX Human ICOS ASYQP
SDP
Catalog No. 5000078400
Encompass
The U-PLEX Human ICOS Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of human Inducible T-cell COStimulator (ICOS) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay can be used to monitor ICOS levels during immunotherapy or measure ICOS on activated T cells With minimal hands-on time it enables efficient and reproducible analysis of ICOS biomarkers
Catalog No. 50-000-78400 Supplier Meso Scale Discovery Supplier No. K151AHBK21
The U-PLEX Human ICOS Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of human Inducible T-cell COStimulator (ICOS) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay can be used to monitor ICOS levels during immunotherapy or measure ICOS on activated T cells With minimal hands-on time it enables efficient and reproducible analysis of ICOS biomarkers