Learn More
Meso Scale Discovery U-PLEX Human ICOS-L/B7-H2 ASY
SDP
Supplier:  Meso Scale Discovery K151AHJK1
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the efficient analysis of ICOS and B7-H2 which play important roles in immune regulation
Catalog No. 50-000-81591