Learn More
Meso Scale Discovery U-PLEX Human IFN- ASY
SDP
Catalog No. 5000082304
Encompass
The Meso Scale Discovery U-PLEX Human IFN gamma Assay is an electrochemiluminescent immunoassay for measuring human interferon gamma (IFN-) in various samples It uses MSD s U-PLEX technology for high sensitivity wide dynamic range and quick results requiring a small sample volume with a detection limit of 0 04 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with an MSD SULFO-TAG and calibrators for a standard curve Results are read on MSD s SECTOR Imager aiding research in immunology oncology and vaccine development
Catalog No. 50-000-82304 Supplier Meso Scale Discovery Supplier No. K151TTK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human IFN gamma Assay is an electrochemiluminescent immunoassay for measuring human interferon gamma (IFN-) in various samples It uses MSD s U-PLEX technology for high sensitivity wide dynamic range and quick results requiring a small sample volume with a detection limit of 0 04 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with an MSD SULFO-TAG and calibrators for a standard curve Results are read on MSD s SECTOR Imager aiding research in immunology oncology and vaccine development