Learn More
Meso Scale Discovery U-PLEX Human IFN ASYQP
SDP
Catalog No. 5000079685
Encompass
The U-PLEX Human IFN Assay (K151TTK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interferon-gamma in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and read buffer for streamlined workflow Designed for research applications in immunology inflammation studies and biomarker discovery it provides reliable quantification of this critical cytokine involved in immune responses and inflammatory processes
Catalog No. 50-000-79685 Supplier Meso Scale Discovery Supplier No. K151TTK22
The U-PLEX Human IFN Assay (K151TTK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interferon-gamma in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and read buffer for streamlined workflow Designed for research applications in immunology inflammation studies and biomarker discovery it provides reliable quantification of this critical cytokine involved in immune responses and inflammatory processes