Learn More
Meso Scale Discovery U-PLEX Human IFN-γ Assay, 384-well plate, 1.7 pg/mL LLOD, 1.7-17,000 pg/mL dynamic range, Mouse Monoclonal antibodies, For Research Use Only
SDP
Catalog No. NC2211631
Encompass
U-PLEX Human IFN-γ Assay, 384-well plate, 1.7 pg/mL LLOD, 1.7-17,000 pg/mL dynamic range, Mouse Monoclonal antibodies, For Research Use Only
This item is not returnable. View return policy
Catalog No. NC2211631 Supplier Meso Scale Discovery Supplier No. K251TTK2
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human IFN-γ Assay, 384-well plate, 1.7 pg/mL LLOD, 1.7-17,000 pg/mL dynamic range, Mouse Monoclonal antibodies, For Research Use Only