Learn More
Meso Scale Discovery U-PLEX Human IFN2a ASYQP
SDP
Catalog No. 5000082362
Encompass
The U-PLEX Human IFN2a Assay (K151VHK-2) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure human interferon alpha-2a levels in biological samples This multiplex-ready assay utilizes MSD s proprietary U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying immune responses and cytokine signaling pathways
Catalog No. 50-000-82362 Supplier Meso Scale Discovery Supplier No. K151VHK21
The U-PLEX Human IFN2a Assay (K151VHK-2) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure human interferon alpha-2a levels in biological samples This multiplex-ready assay utilizes MSD s proprietary U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying immune responses and cytokine signaling pathways